Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftr56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003200557.5 | XP_003200605.1 | 5 - 6 | 1227 | 81 | |
GenBank mRNA | AM941319.1 | - | 5 - 6 | 921 | 81 | |
AM941325.1 | - | 5 - 6 | 921 | 81 | ||
AM941336.1 | - | 4 - 5 | 1054 | 81 | ||
AM941360.1 | - | 5 - 6 | 921 | 81 | ||
AM941361.1 | - | 4 - 5 | 1054 | 81 | ||
CF999928.1 | - | 2 - 3 | 191 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Danio rerio Probe spans exons |