Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045092.1 | NP_001038557.1 | 4 - 5 | 1077 | 79 | |
XM_021469350.1 | XP_021325025.1 | 4 - 5 | 79 | |||
XM_021469351.1 | XP_021325026.1 | 3 - 4 | 79 | |||
XM_021469352.1 | XP_021325027.1 | 4 - 5 | 79 | |||
GenBank mRNA | BC163262.1 | - | 4 - 5 | 79 | ||
DQ241789.1 | - | 4 - 5 | 1077 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |