Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lman2lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076733.1 | NP_001070201.1 | 5 - 6 | 701 | 85 | |
GenBank mRNA | BC124334.1 | - | 5 - 6 | 701 | 85 | |
BC165327.1 | - | 5 - 6 | 710 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |