Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem88b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077144.2 | NP_001070612.2 | 2 - 3 | 624 | 66 | |
XM_005172208.4 | XP_005172265.1 | 2 - 3 | 789 | 66 | ||
GenBank mRNA | BC091536.1 | - | 3 - 4 | 516 | 66 | |
BC124314.1 | - | 2 - 3 | 605 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |