Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077173.1 | NP_001070641.1 | 4 - 5 | 328 | 75 | |
XM_005158223.4 | XP_005158280.1 | 3 - 4 | 501 | 75 | ||
GenBank mRNA | BC124210.1 | - | 4 - 5 | 328 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |