Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077389.1 | NP_001070857.1 | 1 - 2 | 242 | 106 | |
GenBank mRNA | BC124667.1 | - | 1 - 2 | 242 | 106 | |
BC165544.1 | - | 1 - 2 | 114 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |