Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077389.1 | NP_001070857.1 | 5 - 6 | 690 | 66 | |
GenBank mRNA | BC124667.1 | - | 5 - 6 | 690 | 66 | |
BC165544.1 | - | 5 - 6 | 562 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |