Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001346412.1 | NP_001333341.1 | 2 - 3 | 416 | 76 | |
NR_144433.1 | 2 - 3 | 416 | 76 | |||
GenBank mRNA | BC162967.1 | - | 2 - 3 | 282 | 76 | |
DQ489726.1 | - | 2 - 3 | 416 | 76 | ||
DQ489727.1 | - | 2 - 3 | 416 | 76 | ||
DQ489728.1 | - | 2 - 3 | 416 | 76 | ||
DQ489729.1 | - | 2 - 3 | 416 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |