Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_144433.1 | 5 - 6 | 886 | 123 | ||
GenBank mRNA | BC162967.1 | - | 5 - 6 | 752 | 123 | |
DQ489727.1 | - | 5 - 6 | 886 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |