Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079679.3 | NP_001073147.1 | 5 - 6 | 549 | 89 | |
XM_005168711.4 | XP_005168768.1 | 5 - 6 | 590 | 89 | ||
XM_005168712.4 | XP_005168769.1 | 5 - 6 | 547 | 89 | ||
XM_005168713.4 | XP_005168770.1 | 5 - 6 | 590 | 89 | ||
GenBank mRNA | BC124539.1 | - | 4 - 5 | 501 | 89 | |
DT063943.1 | - | 5 - 6 | 532 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |