Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079679.3 | NP_001073147.1 | 6 - 7 | 611 | 139 | |
XM_005168711.4 | XP_005168768.1 | 6 - 7 | 652 | 139 | ||
XM_005168712.4 | XP_005168769.1 | 6 - 7 | 609 | 139 | ||
XM_005168713.4 | XP_005168770.1 | 6 - 7 | 652 | 139 | ||
GenBank mRNA | BC124539.1 | - | 5 - 6 | 563 | 139 | |
DT063943.1 | - | 6 - 7 | 594 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |