Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrxn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079970.1 | NP_001073439.1 | 21 - 22 | 4508 | 94 | |
XM_005161353.4 | XP_005161410.1 | 4 - 5 | 1977 | 94 | ||
XM_009295476.3 | XP_009293751.1 | 21 - 22 | 4709 | 94 | ||
XM_009295477.3 | XP_009293752.1 | 20 - 21 | 4685 | 94 | ||
XM_009295478.3 | XP_009293753.1 | 20 - 21 | 4691 | 94 | ||
XM_009295479.3 | XP_009293754.1 | 4 - 5 | 1972 | 94 | ||
XM_021468849.1 | XP_021324524.1 | 21 - 22 | 4733 | 94 | ||
XM_021468850.1 | XP_021324525.1 | 21 - 22 | 4499 | 94 | ||
XM_021468851.1 | XP_021324526.1 | 20 - 21 | 4706 | 94 | ||
XM_021468852.1 | XP_021324527.1 | 19 - 20 | 3786 | 94 | ||
XM_021468853.1 | XP_021324528.1 | 19 - 20 | 4664 | 94 | ||
XM_021468855.1 | XP_021324530.1 | 21 - 22 | 4739 | 94 | ||
GenBank mRNA | DQ641428.1 | - | 21 - 22 | 4508 | 94 | |
DQ641429.1 | - | 4 - 5 | 1911 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |