Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080076.1 | NP_001073545.1 | 3 - 4 | 429 | 71 | |
XM_005167226.4 | XP_005167283.1 | 3 - 4 | 493 | 71 | ||
GenBank mRNA | AM901599.1 | - | 3 - 4 | 460 | 71 | |
BC128871.1 | - | 3 - 4 | 429 | 71 | ||
KM655634.1 | - | 3 - 4 | 215 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |