Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082809.1 | NP_001076278.1 | 1 - 2 | 166 | 126 | |
GenBank mRNA | BC133925.1 | - | 1 - 2 | 119 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |