Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npvf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082949.1 | NP_001076418.1 | 1 - 2 | 155 | 120 | |
GenBank mRNA | AB193139.1 | - | 1 - 2 | 155 | 120 | |
BC163153.1 | - | 1 - 2 | 155 | 120 | ||
BC163155.1 | - | 1 - 2 | 155 | 120 | ||
EF547661.1 | - | 1 - 2 | 153 | 120 | ||
GU290218.1 | - | 1 - 2 | 136 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |