Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zcchc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089339.2 | NP_001082808.2 | 2 - 3 | 217 | 123 | |
GenBank mRNA | BC134199.1 | - | 2 - 3 | 209 | 123 | |
EH555889.1 | - | 2 - 3 | 252 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |