Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104649.2 | NP_001098119.2 | 10 - 11 | 1109 | 85 | |
XM_021471707.1 | XP_021327382.1 | 9 - 10 | 1082 | 85 | ||
XM_021471709.1 | XP_021327384.1 | 8 - 9 | 872 | 85 | ||
XM_021471716.1 | XP_021327391.1 | 10 - 11 | 1171 | 85 | ||
GenBank mRNA | BC163912.1 | - | 10 - 11 | 1131 | 85 | |
EF653429.1 | - | 10 - 11 | 1191 | 85 | ||
EU124003.1 | - | 10 - 11 | 1123 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |