Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104649.2 | NP_001098119.2 | 8 - 9 | 891 | 70 | |
XM_021471707.1 | XP_021327382.1 | 7 - 8 | 864 | 70 | ||
XM_021471709.1 | XP_021327384.1 | 6 - 7 | 654 | 70 | ||
XM_021471716.1 | XP_021327391.1 | 8 - 9 | 953 | 70 | ||
GenBank mRNA | BC163912.1 | - | 8 - 9 | 913 | 70 | |
EF653429.1 | - | 8 - 9 | 973 | 70 | ||
EU124003.1 | - | 8 - 9 | 905 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |