Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nipsnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131033.2 | NP_571108.2 | 1 - 2 | 374 | 86 | |
GenBank mRNA | AJ249796.1 | - | 1 - 2 | 86 | ||
BC122249.1 | - | 1 - 2 | 374 | 86 | ||
EH513303.1 | - | 1 - 2 | 133 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |