Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nipsnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131033.2 | NP_571108.2 | 3 - 4 | 547 | 130 | |
GenBank mRNA | AJ249796.1 | - | 3 - 4 | 247 | 130 | |
BC122249.1 | - | 3 - 4 | 547 | 130 | ||
EH513303.1 | - | 3 - 4 | 306 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |