Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131114.1 | NP_571189.1 | 2 - 3 | 449 | 78 | |
XM_005171553.4 | XP_005171610.1 | 2 - 3 | 920 | 78 | ||
GenBank mRNA | BC071289.1 | - | 2 - 3 | 394 | 78 | |
BC151893.1 | - | 2 - 3 | 395 | 78 | ||
X71845.1 | - | 449 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |