Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271820.1 | NP_001258749.1 | 3 - 4 | 278 | 97 | |
NM_001328587.1 | NP_001315516.1 | 3 - 4 | 275 | 97 | ||
NM_131327.2 | NP_571402.1 | 3 - 4 | 275 | 97 | ||
XM_005165101.4 | XP_005165158.1 | 3 - 4 | 137 | 97 | ||
GenBank mRNA | AF365873.1 | - | 3 - 4 | 245 | 97 | |
CT619016.1 | - | 3 - 4 | 236 | 97 | ||
CT635558.2 | - | 3 - 4 | 244 | 97 | ||
EH435782.1 | - | 3 - 4 | 282 | 97 | ||
U60804.1 | - | 3 - 4 | 147 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |