Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpeb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131427.2 | NP_571502.2 | 4 - 5 | 625 | 71 | |
XM_021477394.1 | XP_021333069.1 | 4 - 5 | 654 | 71 | ||
GenBank mRNA | AB011680.1 | - | 3 - 4 | 589 | 71 | |
AF076918.1 | - | 4 - 5 | 604 | 71 | ||
BC045467.1 | - | 4 - 5 | 623 | 71 | ||
BC164038.1 | - | 4 - 5 | 623 | 71 | ||
EH503358.1 | - | 4 - 5 | 660 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |