Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: notch1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131441.1 | NP_571516.1 | 28 - 29 | 5592 | 64 | |
GenBank mRNA | X69088.1 | - | 5592 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |