Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244600.1 | NP_001231529.1 | 6 - 7 | 1354 | 106 | |
NM_001244601.1 | NP_001231530.1 | 6 - 7 | 1228 | 106 | ||
NM_131453.3 | NP_571528.2 | 6 - 7 | 989 | 106 | ||
XM_005166786.4 | XP_005166843.1 | 5 - 6 | 1266 | 106 | ||
XM_017357272.2 | XP_017212761.1 | 5 - 6 | 1105 | 106 | ||
XM_021478178.1 | XP_021333853.1 | 6 - 7 | 1343 | 106 | ||
GenBank mRNA | BC065965.1 | - | 6 - 7 | 989 | 106 | |
U17602.1 | - | 5 - 6 | 835 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |