Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cry1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131790.4 | NP_571865.4 | 7 - 8 | 1171 | 96 | |
GenBank mRNA | AB042249.1 | - | 6 - 7 | 826 | 96 | |
BC044558.1 | - | 7 - 8 | 1171 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |