Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldoaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194377.2 | NP_919358.2 | 4 - 5 | 652 | 94 | |
XM_005164116.4 | XP_005164173.1 | 4 - 5 | 698 | 94 | ||
GenBank mRNA | AF533645.1 | - | 4 - 5 | 94 | ||
AY394966.1 | - | 4 - 5 | 652 | 94 | ||
BC044379.1 | - | 4 - 5 | 631 | 94 | ||
BC065320.1 | - | 4 - 5 | 602 | 94 | ||
BC164192.1 | - | 3 - 4 | 571 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |