Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197934.1 | NP_922916.1 | 5 - 6 | 1147 | 93 | |
GenBank mRNA | AF533661.1 | - | 5 - 6 | 1147 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |