Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tardbpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001362353.1 | NP_001349282.1 | 4 - 5 | 1111 | 58 | |
NM_198364.2 | NP_938178.1 | 4 - 5 | 1126 | 58 | ||
XM_021469761.1 | XP_021325436.1 | 4 - 5 | 1179 | 58 | ||
XM_021469762.1 | XP_021325437.1 | 4 - 5 | 1194 | 58 | ||
GenBank mRNA | BC044405.1 | - | 4 - 5 | 995 | 58 | |
BC066430.1 | - | 4 - 5 | 1111 | 58 | ||
BC164253.1 | - | 4 - 5 | 766 | 58 | ||
EH537260.1 | - | 4 - 5 | 599 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |