Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198813.1 | NP_942108.1 | 1 | 3149 | 141 | |
XM_009296582.3 | XP_009294857.1 | 2 | 3487 | 141 | ||
GenBank mRNA | AY311506.1 | - | 1 | 3149 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |