Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199591.2 | NP_955885.1 | 4 - 5 | 744 | 72 | |
XM_005160454.3 | XP_005160511.1 | 5 - 6 | 868 | 72 | ||
GenBank mRNA | AY577010.1 | - | 5 - 6 | 811 | 72 | |
BC049502.1 | - | 3 - 4 | 383 | 72 | ||
BC066614.1 | - | 4 - 5 | 744 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |