Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199671.1 | NP_955965.1 | 3 - 4 | 288 | 99 | |
GenBank mRNA | BC064308.1 | - | 3 - 4 | 288 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |