Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx39aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328342.1 | NP_001315271.1 | 9 - 10 | 1351 | 71 | |
GenBank mRNA | BC042330.1 | - | 10 - 11 | 1265 | 71 | |
BC071505.1 | - | 9 - 10 | 1353 | 71 | ||
EH559791.1 | - | 1 - 2 | 256 | 71 | ||
EH570750.1 | - | 1 - 2 | 279 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |