Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199784.1 | NP_956078.1 | 5 - 6 | 674 | 63 | |
GenBank mRNA | BC056728.1 | - | 5 - 6 | 674 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |