Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199819.2 | NP_956113.1 | 5 - 6 | 689 | 99 | |
GenBank mRNA | BC044467.1 | - | 5 - 6 | 693 | 99 | |
BC064289.1 | - | 5 - 6 | 669 | 99 | ||
EH432914.1 | - | 5 - 6 | 721 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |