Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200034.1 | NP_956328.1 | 1 - 2 | 193 | 79 | |
GenBank mRNA | BC049018.1 | - | 1 - 2 | 193 | 79 | |
BC071411.1 | - | 1 - 2 | 176 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |