Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pigm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200390.1 | NP_956684.1 | 2 - 3 | 716 | 76 | |
GenBank mRNA | BC053299.1 | - | 2 - 3 | 716 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |