Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200419.1 | NP_956713.1 | 5 - 6 | 1066 | 127 | |
XM_021479120.1 | XP_021334795.1 | 4 - 5 | 1132 | 127 | ||
GenBank mRNA | BC054637.1 | - | 5 - 6 | 1066 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |