Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkain1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200545.1 | NP_956839.1 | 5 - 6 | 871 | 83 | |
GenBank mRNA | BC056509.1 | - | 5 - 6 | 871 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |