Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: macrod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200549.1 | NP_956843.1 | 18 - 19 | 1304 | 62 | |
XM_017358720.2 | XP_017214209.1 | 19 - 20 | 1466 | 62 | ||
GenBank mRNA | BC056529.1 | - | 18 - 19 | 1304 | 62 | |
BC152658.1 | - | 18 - 19 | 1385 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |