Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: surf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201065.1 | NP_957359.1 | 3 - 4 | 462 | 79 | |
XM_005171804.4 | XP_005171861.1 | 3 - 4 | 494 | 79 | ||
GenBank mRNA | AY648750.1 | - | 3 - 4 | 444 | 79 | |
BC053193.1 | - | 3 - 4 | 462 | 79 | ||
BC164187.1 | - | 2 - 3 | 329 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |