Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: id2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201291.1 | NP_958448.1 | 1 - 2 | 417 | 127 | |
GenBank mRNA | AY398363.1 | - | 1 - 2 | 440 | 127 | |
BC056303.1 | - | 1 - 2 | 417 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |