Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dldh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201506.1 | NP_958914.1 | 7 - 8 | 639 | 118 | |
GenBank mRNA | AY391421.1 | - | 7 - 8 | 590 | 118 | |
BC044432.1 | - | 7 - 8 | 639 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |