Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec61a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201577.1 | NP_963871.1 | 7 - 8 | 683 | 74 | |
GenBank mRNA | AY034614.1 | - | 7 - 8 | 683 | 74 | |
BC048881.1 | - | 7 - 8 | 684 | 74 | ||
BC165352.1 | - | 6 - 7 | 649 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |