Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacna1da | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203484.1 | NP_982351.1 | 45 - 46 | 5738 | 76 | |
XM_021479576.1 | XP_021335251.1 | 46 - 47 | 5959 | 76 | ||
XM_021479577.1 | XP_021335252.1 | 46 - 47 | 5959 | 76 | ||
XM_021479578.1 | XP_021335253.1 | 46 - 47 | 5956 | 76 | ||
XM_021479579.1 | XP_021335254.1 | 45 - 46 | 5932 | 76 | ||
XM_021479580.1 | XP_021335255.1 | 45 - 46 | 5925 | 76 | ||
XM_021479581.1 | XP_021335256.1 | 45 - 46 | 5883 | 76 | ||
XM_021479582.1 | XP_021335257.1 | 44 - 45 | 5578 | 76 | ||
XM_021479583.1 | XP_021335258.1 | 45 - 46 | 5842 | 76 | ||
XM_021479584.1 | XP_021335259.1 | 44 - 45 | 5850 | 76 | ||
GenBank mRNA | AY528224.1 | - | 45 - 46 | 5738 | 76 | |
BC163679.1 | - | 44 - 45 | 5578 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |