Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328492.1 | NP_001315421.1 | 6 - 7 | 988 | 84 | |
NM_001328493.1 | NP_001315422.1 | 5 - 6 | 817 | 84 | ||
NM_205702.2 | NP_991265.1 | 4 - 5 | 516 | 84 | ||
GenBank mRNA | BC065985.1 | - | 4 - 5 | 516 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |