Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: per1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212439.2 | NP_997604.2 | 2 - 3 | 987 | 73 | |
GenBank mRNA | AY597250.1 | - | 2 - 3 | 987 | 73 | |
BC163718.1 | - | 1 - 2 | 532 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |