Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: decr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213321.1 | NP_998486.1 | 5 - 6 | 470 | 67 | |
XM_005162880.4 | XP_005162937.1 | 5 - 6 | 500 | 67 | ||
GenBank mRNA | BC068332.1 | - | 5 - 6 | 470 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |