Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214730.1 | NP_999895.1 | 4 - 5 | 972 | 87 | |
XM_005172174.4 | XP_005172231.1 | 4 - 5 | 1017 | 87 | ||
XM_005172175.4 | XP_005172232.1 | 4 - 5 | 1020 | 87 | ||
GenBank mRNA | BC065964.1 | - | 4 - 5 | 972 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |