Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001730.2 | NP_001001730.2 | 3 - 4 | 511 | 89 | |
GenBank mRNA | AY496939.1 | - | 3 - 4 | 420 | 89 | |
BC116552.1 | - | 3 - 4 | 511 | 89 | ||
BC164033.1 | - | 3 - 4 | 454 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |